4-Chloro-2-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21301G100 AS21301G25
ACTIAS21301G100EA 101 GBP
ACTIAS21301G100 ACTIAS21301G25
4-Chloro-2-nitrobenzonitrile
4-Chloro-2-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
MDL Number: MFCD00027398
CAS Number: 34662-32-3
EINECS: 252-133-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR