3,5-Difluoro-DL-phenylalanine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004859-1G 004859-2.5G
FLUO004859-1GEA 0 GBP
FLUO004859-1G FLUO004859-2.5G
3,5-Difluoro-DL-phenylalanine
3,5-Difluoro-DL-phenylalanine
Formula: C₉H₉F₂NO₂
MW: 201.1701 g/mol
MDL Number: MFCD00061308
CAS Number: 32133-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR