2,5-Difluoro-DL-phenylalanine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-3-(2,5-difluorophenyl)propanoic acid, H-DL-Phe(2,5-DiF)-OH

004857-500MG 004857-1G
FLUO004857-500MGEA 0 GBP
FLUO004857-500MG FLUO004857-1G
2,5-Difluoro-DL-phenylalanine
2,5-Difluoro-DL-phenylalanine
Formula: C₉H₉F₂NO₂
MW: 201.170 g/mol
MDL Number: MFCD00061303
CAS Number: 32133-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR