1,4-Dibromo-2,5-bis(bromomethyl)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0536-5G OT-0536-1G
COBBOT-0536-5GEA 0 GBP
COBBOT-0536-5G COBBOT-0536-1G
1,4-Dibromo-2,5-bis(bromomethyl)benzene
1,4-Dibromo-2,5-bis(bromomethyl)benzene
Formula: C₈H₆Br₄
MW: 421.7500 g/mol
MDL Number: MFCD03411745
CAS Number: 35335-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR