1,3-Dibromo-5-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0339-5G OT-0339-25G OT-0339-100G
COBBOT-0339-5GEA 0 GBP
COBBOT-0339-5G COBBOT-0339-25G COBBOT-0339-100G
1,3-Dibromo-5-(trifluoromethoxy)benzene
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formula: C₇H₃Br₂F₃O
MW: 319.90 g/mol
MDL Number: MFCD00075280
CAS Number: 207226-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR