3-Fluoro-4-methylbenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005796-1G
005796-5G
005796-10G
FLUO005796-1GEA
0
GBP
FLUO005796-1G
FLUO005796-5G
FLUO005796-10G
3-Fluoro-4-methylbenzylamine
3-Fluoro-4-methylbenzylamine
Formula:
C₈H₁₀FN MW: 139.17 g/mol |
MDL Number:
MFCD01631531 CAS Number: 261951-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...