Allyl methyl sulphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Allyl(methyl)sulphane
M-Clarity™ quality level = MQ200
A34201-25G
A34201-5G
SIALA34201-25GEA
113
GBP
SIALA34201-25G
SIALA34201-5G
Allyl methyl sulphide, Sigma-Aldrich®
Allyl methyl sulphide
Formula:
C₄H₈S MW: 88.17 g/mol |
MDL Number:
MFCD00008657 CAS Number: 10152-76-8 EINECS: 233-422-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...