4-Chloro-8-methoxyquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21916G1 AS21916G5
ACTIAS21916G1EA 0 GBP
ACTIAS21916G1 ACTIAS21916G5
4-Chloro-8-methoxyquinoline
4-Chloro-8-methoxyquinoline
Formula: C₁₀H₈ClNO
MW: 193.6296 g/mol
MDL Number: MFCD00033679
CAS Number: 16778-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR