3-Benzylcarbamoyl-5-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Benzylcarbamoyl)-5-fluorophenylboronic acid

BB-2859-5G BB-2859-1G
COBBBB-2859-5GEA 0 GBP
COBBBB-2859-5G COBBBB-2859-1G
3-Benzylcarbamoyl-5-fluorophenylboronic acid
3-Benzylcarbamoyl-5-fluorophenylboronic acid
Formula: C₁₄H₁₃BFNO₃
MDL Number: MFCD08235058
CAS Number: 874219-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR