3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Bromo-2-((3-fluorobenzyl)oxy)-5-methylphenyl)boronic acid

BB-8053-1G BB-8053-250M
COBBBB-8053-1GEA 0 GBP
COBBBB-8053-1G COBBBB-8053-250M
3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid
3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid
Formula: C₁₄H₁₃BBrFO₃
MW: 338.9700 g/mol
MDL Number: MFCD06411359
CAS Number: 849062-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR