2-(Benzylthio)ethanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxyethyl Benzyl Sulphide, 2-(Benzylthio)ethan-1-ol

003733-25G 003733-100G
FLUO003733-25GEA 0 GBP
FLUO003733-25G FLUO003733-100G
2-(Benzylthio)ethanol
2-(Benzylthio)ethanol
Formula: C₉H₁₂OS
MW: 168.25 g/mol
MDL Number: MFCD00014039
CAS Number: 3878-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR