4-Hydroxy-2-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8401-1G BB-8401-250M
COBBBB-8401-1GEA 0 GBP
COBBBB-8401-1G COBBBB-8401-250M
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₃
MDL Number: MFCD03095345
CAS Number: 943918-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR