4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxy-2-(trifluoromethyl)phenylboronic acid, pinacol ester

PN-8401-1G PN-8401-250M
COBBPN-8401-1GEA 0 GBP
COBBPN-8401-1G COBBPN-8401-250M
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₃
MDL Number: MFCD15143616
CAS Number: 1029439-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR