3-Chloro-N-4-dimethylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2874-5G
HC-2874-1G
COBBHC-2874-5GEA
0
GBP
COBBHC-2874-5G
COBBHC-2874-1G
3-Chloro-N-4-dimethylaniline hydrochloride
3-Chloro-N,4-dimethylaniline hydrochloride
Formula:
C₈H₁₁Cl₂N |
MDL Number:
MFCD12756444 CAS Number: 7745-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...