3'-Bromo-5'-chloro-2'-hydroxyacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3-Bromo-5-chloro-2-hydroxyphenyl)ethanone, 5-Chloro-3-bromo-2-hydroxyacetophenone

AS25308G25 AS25308G5
ACTIAS25308G25EA 95 GBP
ACTIAS25308G25 ACTIAS25308G5
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD00857068
CAS Number: 59443-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR