2-(Methylsulphonyl)phenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0401-1G OT-0401-5G
COBBOT-0401-1GEA 0 GBP
COBBOT-0401-1G COBBOT-0401-5G
2-(Methylsulphonyl)phenol
2-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
MDL Number: MFCD00127516
CAS Number: 27489-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR