2-Fluoro-5-(methylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid, 2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

BB-3335-1G BB-3335-5G
COBBBB-3335-1GEA 0 GBP
COBBBB-3335-1G COBBBB-3335-5G
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BFNO₃
MW: 196.97 g/mol
MDL Number: MFCD08436028
CAS Number: 874289-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR