4-Aminophenylboronic acid hydrochloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

708887-250MG 708887-1G
SIAL708887-250MGEA 178 GBP
SIAL708887-250MG SIAL708887-1G
4-Aminophenylboronic acid hydrochloride, Sigma-Aldrich®
4-Aminophenylboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR