2-Aminophenylboronic acid hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2228-1G BB-2228-5G
COBBBB-2228-1GEA 0 GBP
COBBBB-2228-1G COBBBB-2228-5G
2-Aminophenylboronic acid hydrochloride
2-Aminophenylboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MW: 173.4052 g/mol
MDL Number: MFCD02258096
CAS Number: 863753-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR