1-Methyl-5-indoleboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methylindole-5-boronic acid, N-Methylindole-5-boronic acid

BB-3928-5G BB-3928-1G
COBBBB-3928-5GEA 0 GBP
COBBBB-3928-5G COBBBB-3928-1G
1-Methyl-5-indoleboronic acid
1-Methyl-5-indoleboronic acid
Formula: C₉H₁₀BNO₂
MW: 174.9900 g/mol
MDL Number: MFCD03412070
CAS Number: 192182-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR