1-Methyl-2-indoleboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1-Methyl-1H-indol-2-yl)boronic acid

BB-2401-5G BB-2401-1G
COBBBB-2401-5GEA 0 GBP
COBBBB-2401-5G COBBBB-2401-1G
1-Methyl-2-indoleboronic acid
1-Methyl-2-indoleboronic acid
Formula: C₉H₁₀BNO₂
MW: 174.99 g/mol
MDL Number: MFCD01114668
CAS Number: 191162-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR