4-Indoleboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

AS12914G1 AS12914G25 AS12914G5
ACTIAS12914G1EA 41 GBP
ACTIAS12914G1 ACTIAS12914G25 ACTIAS12914G5
4-Indoleboronic acid pinacol ester
4-Indoleboronic acid pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR