Dibenzyl phosphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

D36550-25G D36550-5G
SIALD36550-25GEA 103 GBP
SIALD36550-25G SIALD36550-5G
Dibenzyl phosphate, Sigma-Aldrich®
Dibenzyl phosphate
Formula: C₁₄H₁₅O₄P
MDL Number: MFCD00004775
CAS Number: 1623-08-1
EINECS: 216-602-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR