Ebselen 99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

ACRO300572500EA 703 GBP
ACRO300572500
Ebselen 99%
Ebselen
Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
Melting Pt: 178…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 184 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR