4-(1H-Pyrazol-1-yl)aniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Pyrazol-1-yl-phenylamine

AS20709G25 AS20709G5
ACTIAS20709G25EA 0 GBP
ACTIAS20709G25 ACTIAS20709G5
4-(1H-Pyrazol-1-yl)aniline
4-(1H-Pyrazol-1-yl)aniline
Formula: C₉H₉N₃
MW: 159.1879 g/mol
MDL Number: MFCD00626298
CAS Number: 17635-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR