1-Benzyl-4-hydroxypiperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Benzylpiperidin-4-ol

AS20529G100 AS20529G25
ACTIAS20529G100EA 0 GBP
ACTIAS20529G100 ACTIAS20529G25
1-Benzyl-4-hydroxypiperidine
1-Benzyl-4-hydroxypiperidine
Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
MDL Number: MFCD00006503
CAS Number: 4727-72-4
EINECS: 225-226-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR