Methyl-5-bromosalicylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-bromo-2-hydroxybenzoate

AS23434G100 AS23434G25
ACTIAS23434G100EA 179 GBP
ACTIAS23434G100 ACTIAS23434G25
Methyl-5-bromosalicylate
Methyl 5-bromosalicylate
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00016460
CAS Number: 4068-76-2
EINECS: 223-776-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR