3-Fluoro-2-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32676G1 AS32676G5
ACTIAS32676G1EA 0 GBP
ACTIAS32676G1 ACTIAS32676G5
3-Fluoro-2-nitrotoluene
3-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD08458093
CAS Number: 3013-27-2
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR