5-Formylsalicylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Formyl-2-hydroxybenzoic acid

AS20753G100 AS20753G25
ACTIAS20753G100EA 124 GBP
ACTIAS20753G100 ACTIAS20753G25
5-Formylsalicylic acid
5-Formylsalicylic acid
Formula: C₈H₆O₄
MW: 166.1308 g/mol
MDL Number: MFCD00006945
CAS Number: 616-76-2
EINECS: 210-492-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR