3,6-Dichloropicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,6-Dichloropyridine-2-carboxylic acid

AS21924G100 AS21924G25
ACTIAS21924G100EA 0 GBP
ACTIAS21924G100 ACTIAS21924G25
3,6-Dichloropicolinic acid
3,6-Dichloropicolinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192.00 g/mol
MDL Number: MFCD00078655
CAS Number: 1702-17-6
EINECS: 216-935-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR