6-Bromo-4-chloroquinazoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2437G1
AS2437G5
ACTIAS2437G1EA
0
GBP
ACTIAS2437G1
ACTIAS2437G5
6-Bromo-4-chloroquinazoline
6-Bromo-4-chloroquinazoline
Formula:
C₈H₄BrClN₂ MW: 243.4878 g/mol |
MDL Number:
MFCD01862191 CAS Number: 38267-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...