6-Morpholinonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20710G1 AS20710G5
ACTIAS20710G1EA 112 GBP
ACTIAS20710G1 ACTIAS20710G5
6-Morpholinonicotinic acid
6-Morpholinonicotinic acid
Formula: C₁₀H₁₂N₂O₃
MW: 208.213 g/mol
MDL Number: MFCD02682034
CAS Number: 120800-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR