3-Phenoxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24068G25 AS24068G5
ACTIAS24068G25EA 375 GBP
ACTIAS24068G25 ACTIAS24068G5
3-Phenoxybenzonitrile
3-Phenoxybenzonitrile
Formula: C₁₃H₉NO
MDL Number: MFCD05669318
CAS Number: 50789-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR