2-Chloropyrimidine-5-carbonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS31780G1 AS31780G5
ACTIAS31780G1EA 0 GBP
ACTIAS31780G1 ACTIAS31780G5
2-Chloropyrimidine-5-carbonitrile
2-Chloropyrimidine-5-carbonitrile
Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
MDL Number: MFCD10697145
CAS Number: 1753-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR