Men's shirt, Legacy

Supplier: Fristads Kansas Group
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Legacy
113-6671EA 39.2 GBP
113-6671 113-6690 113-6670 113-6679 113-6659 113-6678 113-6661 113-6683 113-6660 113-6682 113-6685 113-6663 113-6684 113-6662 113-6681 113-6680 113-6669 113-6668 113-6665 113-6687 113-6664 113-6686 113-6689 113-6667 113-6688 113-6666
Men's shirt, Legacy
Workwear
This robust shirt manufactured from 100% combed cotton is made for heavy-duty wear. Suitable for industrial washing.

  • Long sleeved
  • Double yoke, reinforced seams
  • Material is shrink-treated
  • Two chest pockets with flap, pencil pocket, mobile phone pocket

Weight: 190 g/m²
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR