(1-Hexadecyl)tributylphosphonium bromide ≥98%

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Hexadecyltri-n-butylphosphonium bromide(n-Hexadecyl)tri-n-butylphosphonium bromideTributyl(hexadecyl)phosphonium bromideTributylhexadecylphosphonium bromide

ABCRAB121580.0010EA 0 GBP
ABCRAB121580.0010
(1-Hexadecyl)tributylphosphonium bromide ≥98%
(1-Hexadecyl)tributylphosphonium bromide
Formula: C₂₈H₆₀BrP
MW: 507.65 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD00011775
CAS Number: 14937-45-2
EINECS: 239-014-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR