3-Fluoro-5-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51589G1
AS51589G5
ACTIAS51589G1EA
0
GBP
ACTIAS51589G1
ACTIAS51589G5
3-Fluoro-5-(tributylstannyl)pyridine
3-Fluoro-5-(tributylstannyl)pyridine
Formula:
C₁₇H₃₀FNSn |
MDL Number:
MFCD13152000 CAS Number: 871942-19-7 UN: 2788 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...