1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

010752-5G 010752-1G
FLUO010752-5GEA 0 GBP
FLUO010752-5G FLUO010752-1G
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₆H₂₁BN₂O₂
MW: 284.16 g/mol
MDL Number: MFCD03789252
CAS Number: 761446-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR