5-Methylsalicylaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxy-5-methylbenzaldehyde

015625-1G 015625-25G
FLUO015625-1GEA 0 GBP
FLUO015625-1G FLUO015625-25G
5-Methylsalicylaldehyde
5-Methylsalicylaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 54 °C
Density: 1.0913 g/cm³ (59 °C)
Storage Temperature: Ambient
MDL Number: MFCD00274297
CAS Number: 613-84-3
EINECS: 210-357-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR