Diethyl fluoromalonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-diethyl 2-fluoropropanedioate

003198-1G 003198-5G
FLUO003198-1GEA 0 GBP
FLUO003198-1G FLUO003198-5G
Diethyl fluoromalonate
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR