3-Iodobenzonitrile
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FLUO147901-5GEA
0
GBP
FLUO147901-25G
FLUO147901-5G
3-Iodobenzonitrile
3-Iodobenzonitrile
Formula:
C₇H₄IN MW: 229.02 g/mol |
MDL Number:
MFCD00079762 CAS Number: 69113-59-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...