4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-(trifluoromethyl)benzene-1-sulphonyl chloride

013073-5G 013073-25G 013073-1G
FLUO013073-5GEA 0 GBP
FLUO013073-5G FLUO013073-25G FLUO013073-1G
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃BrClF₃O₂S
MW: 323.52 g/mol
Melting Pt: 54…58 °C
MDL Number: MFCD03094392
CAS Number: 176225-10-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR