2'-Fluoro-5'-(trifluoromethyl)propiophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006450-1G
006450-5G
006450-10G
FLUO006450-1GEA
0
GBP
FLUO006450-1G
FLUO006450-5G
FLUO006450-10G
2'-Fluoro-5'-(trifluoromethyl)propiophenone
2'-Fluoro-5'-(trifluoromethyl)propiophenone
Formula:
C₁₀H₈F₄O MW: 220.17 g/mol |
MDL Number:
MFCD00061260 CAS Number: 207974-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...