6-Methylindole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Methyl-1H-indole

016244-1G 016244-5G
FLUO016244-1GEA 0 GBP
FLUO016244-1G FLUO016244-5G
6-Methylindole
6-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00005682
CAS Number: 3420-02-8
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR