4-Bromo-2-fluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

016230-1G 016230-5G
FLUO016230-1GEA 0 GBP
FLUO016230-1G FLUO016230-5G
4-Bromo-2-fluoropyridine
4-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04112504
CAS Number: 128071-98-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR