4-tert-Butylphenylhydrazine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-(tert-Butyl)phenyl)hydrazine hydrochloride
M-Clarity™ quality level = MQ100
593877-1G
593877-5G
SIAL593877-1GEA
91.8
GBP
SIAL593877-1G
SIAL593877-5G
4-tert-Butylphenylhydrazine hydrochloride, Sigma-Aldrich®
4-tert-Butylphenylhydrazine hydrochloride
Formula:
C₁₀H₁₇ClN₂ MW: 200.7084 g/mol |
MDL Number:
MFCD00013387 CAS Number: 128231-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...