1,3-Dibromo-5-tert-butylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dibromo-5-(tert-butyl)benzene, 5-(tert-Butyl)-1,3-dibromobenzene

023861-25G 023861-5G
FLUO023861-25GEA 0 GBP
FLUO023861-25G FLUO023861-5G
1,3-Dibromo-5-tert-butylbenzene
1,3-Dibromo-5-tert-butylbenzene
Formula: C₁₀H₁₂Br₂
MW: 292.0100 g/mol
MDL Number: MFCD06657956
CAS Number: 129316-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR