2,5-Difluoro-4-nitrobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010720-1G 010720-5G
FLUO010720-1GEA 0 GBP
FLUO010720-1G FLUO010720-5G
2,5-Difluoro-4-nitrobenzoic acid
2,5-Difluoro-4-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MW: 203.10 g/mol
MDL Number: MFCD03425705
CAS Number: 116465-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR