2-Chloro-6-fluoro-3-methylanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005640-1G 005640-5G
FLUO005640-1GEA 0 GBP
FLUO005640-1G FLUO005640-5G
2-Chloro-6-fluoro-3-methylanisole
2-Chloro-6-fluoro-3-methylanisole
Formula: C₈H₈ClFO
MW: 174.60 g/mol
MDL Number: MFCD01631367
CAS Number: 261762-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR