2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-8606-1G
PN-8606-5G
PN-8606-25G
COBBPN-8606-1GEA
0
GBP
COBBPN-8606-1G
COBBPN-8606-5G
COBBPN-8606-25G
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₇H₁₅BO₃ MW: 158.01 g/mol |
MDL Number:
MFCD03093086 CAS Number: 1195-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...